Would you like to log in again?
PureAmp PCR and ELISA Plate Sealing Films
Supplier: MTC BIO, INC.
P1001-A P1001-E P1001-PCR P1001-QPS P1001-Q
470347-462PK 84.65 USD InStock
470347-462 470347-464 470347-470 470347-466 470347-468
PureAmp PCR and ELISA Plate Sealing Films
Seals Microplate Seals
A versatile sealing film for standard ELISA and PCR 96-well plates.
  • RNase, DNase, DNA, and PCR inhibitor free
  • EZ-Peel adhesive for spill-free removal
  • Inhibits evaporation across all 96 wells (≤5%)
  • Suitable for a wide temperature range
The sealing options in the PureAmp™ line include clear plastic film for end-point PCR, two optically clear films for qPCR, plus one pierceable aluminum membrane for storage and/or PCR. All PureAmp™ sealing products are pre-cut for a perfect fit on the plate every time and are guaranteed to keep evaporation rates under 5% in typical PCR applications. All PureAmp™ tubes, strips and plates are made of medical grade heat-resistant polypropylene. All PureAmp™ products are PCR certified and are guaranteed to be free of RNase, DNase, DNA, pyrogens, endotoxins and PCR inhibitors.
Packaging: Product is packaged in clear zip lock bags of 10 membranes within the standard pack of 100 membranes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty