Would you like to log in again?
Piezoelectric Buzzer
Supplier: United Scientific Supplies
PIEZOBZPK5
470347-894EA 4.9 USD InStock
470347-894
Piezoelectric Buzzer
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Enhance your circuits with sound.
  • Requires low voltage 1 - 3 V
  • Long leads
This piezoelectric buzzer is an electronic sounder with integration structure. Using a 1 – 3 V DC power supply this can be easily integrated into any electrical circuit. The wire leads can be easily connected to other electrical components.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty