Would you like to log in again?
REUZ™ Stainless Steel Balance Spoon
Supplier: United Scientific Supplies
UN1004-175
470347-964EA 4.65 USD InStock
470347-964
REUZ™ Stainless Steel Balance Spoon
Spoons
A slim stainless steel blade for transferring materials.
  • 304 stainless steel
  • Smooth, polished surface
  • Easy to clean
  • Long, slim blade fits easily in containers
  • 175 mm
Manufactured with high-quality 304 stainless steel, the United Scientific REUZ™ ('re-use') balance spoon has a trough-shaped tip to easily pick up and transfer materials. Slim blade with long cavity and a long handle enables the spoon to be used in a variety of containers. Constructed of a single piece of stainless steel with a polished surface to resist contamination. Intended for use with most powders, granules, pastes and liquids.
REUZ™ products promote sustainability in all laboratory environments.
Ordering information: Sold individually.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty