Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SingleShot Buffer Pouches

Supplier: NuSep
DBM-TBE-01 DBM-PBS-01 DBM-SSC-01 DBM-LBM-01 DBM-TGS-01 DBM-LBA-01 DBM-TRG-01 DBM-TAE-01
470348-096EA 62.3 USD InStock
470348-096 470348-100 470348-110 470348-098 470348-104 470348-102 470348-108 470348-106
SingleShot Buffer Pouches
Buffers Biological Buffers
Each pouch contains a convenient, pre-mixed buffer solution; just add water!

  • Quality – molecular biology-grade buffers of highest purity
  • Convenient – pre-measured powder in sealed pouches
  • Fast – dissolve and use right away
  • Standardized – guaranteed reproducible results for your electrophoresis gels and Western blots

NuSep’s SingleShot Buffer pouches are perfect for standardizing your electrophoresis and Western blotting applications. Dissolve a pouch of pre-measured molecular biology-grade chemicals in 500 ml of deionized water to obtain a ready-to-use 1x buffer without any further preparation. Save precious time in the laboratory and generate reproducible results with Avant buffers.

Buffer Selections:
TGS (Tris-Glycine SDS) – Gel Running Buffer-Cat# DBM-TGS-01
PBS (Phosphate Buffer Saline) – Western Blotting-Cat# DBM-PBS-01
TG (Tris-Glycine) – Transfer Buffer-Cat# DBM-TRG-01
TBE (Tris-Boric EDTA Buffer) – DNA/RNA Electrophoresis Buffer-Cat# DBM-TBE-01
TAE (Tris-Acetate EDTA Buffer) – DNA/RNA Electrophoresis Buffer-Cat# DBM-TAE-01
SSC (Saline Sodium Citrate Buffer) – Southern and Northern Hybridization-Cat# DBM-SSC-01

Each packet quickly yields 500 ml of ready-to-use 1x concentration buffer

NuSep also provide LB Agar and LB Media formulations. Buffers are premixed and packaged for your convenience. Just add water and use.

LB Agar (Cat# DBM-LBA-01): Each packet quickly yields 50 ml of ready-to-use 1x concentration working solution.
LB Media (Cat# DBM-LBM-01): Each packet quickly yields 100 ml of ready-to-use 1x concentration working solution.

Delivery information: 20 Buffer Pouches/Box

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty