Would you like to log in again?
Nasco Healthcare® Economy Visual Training Assistant (VTA)
Supplier: NASCO HEALTHCARE INC. SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nasco Healthcare®
100-2161 100-2160B 100-2160
470348-682PK 1077.15 USD InStock
470348-682 470348-686 470348-674
Nasco Healthcare® Economy Visual Training Assistant (VTA)
First Aid Supplies CPR Products
Easy and cost effective.
  • Palpable landmarks
  • Observable chest rise
  • Easy to use VTA
The Econo VTA enables students to train and improve their CPR skills. This CPR trainer has the same look and feel of the lifelike economy Sani-Manikin and is affordable enough for each student to have their own manikin. The simple face shield lung system means no cleaning, disinfecting, or disassembly is required. The features include a head tilt/chin life feature for opening the airway, touch vinyl skin for extended service, and a foam-filled torso for resistance to compressions.
The VTA indication system is easy to understand. A rate light comes on when the correct rate is achieved and a second light comes on to show correct depth with proper release. When the student keeps both lights on, they are delivering quality CPR. Includes three airways and instructions for use.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty