Would you like to log in again?
Crazy Circuits™ Bit Board Rover Kits
Supplier: BROWN DOG GADGETS
Crazy Circuits
K1053-10PK K1053-01PK K1053-05PK K1053-25PK
470348-948EA 4121.3 USD InStock
470348-948 470348-946 470348-928 470348-932
Crazy Circuits™ Bit Board Rover Kits
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Build and program a versatile robot powered by the popular micro:bit system.
  • LEGO™ brick-compatible rover powered by micro:bit
  • Bit board rover kit is designed to teach the fundamentals of robotics and coding - accessible to learners of any age
  • No soldering or additional tools needed
Automation and robotics weave together concepts of programming, circuitry and mechanics to continually revolutionize the way humans get work done in our everyday world. Our brand-new rover kit allows learners to gain experience in these content areas within a familiar brick-building enviroment while using the approachable micro:bit programming interface to control a vehicle that can be operated remotely or autonomously. Build tank and rover frames/grabbers/lifters/sweepers/drivetrains. Program moving/non-moving outcomes. Work with sensing/reacting sequences and control it all in a variety of ways. Affordable and just the right amount of room for customization and challenge, this micro:bit powered kit offers a fun and approachable way to introduce robotics to learners ages 10 and up.
Ordering information: Required, but not included: micro:bit controller.
Delivery information: Kits include (207) building blocks, (2) continuous rotation servos, (2) line following (IR) sensors, (2) sets (of 4) ribbon cables, (1) 270 degree servo, (1) bit board, (1) thumbstick, (1) block-compatible AA battery holder, (1) block-compatible distance sensor, (1) pen holder, (1) Neopixel lighting array.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty