Would you like to log in again?
Motic Swift Line Infinity Corrected Conpound Microscopes with Koehler Illumination
Supplier: MOTIC INSTRUMENTS USA, INC SE
M18B-P M18T-P
470349-298EA 1410.75 USD InStock
470349-298 470349-300
Motic Swift Line Infinity Corrected Conpound Microscopes with Koehler Illumination
Microscopes
The M18 compound microscope series features infinity corrected plan optics, ideal for college laboratories, veterinary and medical applications.
  • Widefield 10×/20 mm eyepieces
  • Binocular/Trinocular viewing head with 30° inclined eyepieces, with diopter and interpupillary adjustment 54 to 76mm
  • Infinity corrected 4× (0.10 N.A.), 10× (0.25 N.A.), 40×R (0.65 N.A.) and 100×R oil immersion (1.25 N.A.) objectives
  • Only the 100×R objective is oil immersion
  • PLAN lenses; all are parfocalled, parcentered and color-coded
  • Quad objective turret is ball-bearing mounted for smooth, precise positioning of objectives. Mounted in reverse position to facilitate ease in changing slides
  • Koehler illumination
  • Coaxial coarse and fine focusing controls, coarse has tension adjustment, fine is graduated
  • Large 5¾×5½" (142×140 mm) stage; built-in, low profile, ball-bearing mounted mechanical specimen holder with low-position coaxial controls
  • 1.25 N.A. Abbe condenser has rack and pinion focusing and iris diaphragm
  • 110 to 220 V switching power supply, unit shipped with American plug
M18B-P is supplied with binocular head, this research level microscope also incorporates Koehler illumination and wide field 20 mm eyepieces for a very crisp image.
M18T-P is supplied with trinocular head, this research level microscope also incorporates Koehler illumination and wide field 20 mm eyepieces for a very crisp image.
Packaging: Head and frame in molded styrofoam container with vinyl dustcover.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty