Would you like to log in again?
ADC® Responder Holsters
Supplier: AMERICAN DIAGNOSTIC CORP CP
ADC®
115BK 105BK 104BKE
470349-472CS 256.39 USD InStock
470349-472 470349-484 470349-478
ADC® Responder Holsters
First Aid Supplies First Aid Kits
Basic tools at hand on your duty belt.
  • Super tough water repellent 600D nylon
  • Sold with basic instruments
  • 1-year warranty
The Responder holster set includes all the essential tools needed by the First Responder. The heavy-duty nylon holster is lined with 8 mm of cushioning foam. Snap open belt loop reinforced with hook and loop adhesive for easy removal and attachment for belts up to 2.5" wide. The 2 deep slide-in pockets in back and 3 slide-in pockets beneath covered flap in front provide storage for the included Kelly forceps, shears, penlight, bandage scissors, knife, f and hook and loop tourniquet.
The responder Junior holsters are a smaller version with room for the included shears, Kelly forceps, and a penlight. These are designed to be worn either horizontally or vertically on a belt.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty