Would you like to log in again?
Class II Laser Pointers
Supplier: United Scientific Supplies
GLPCLS2 RLPCLS2
470349-618EA 38.95 USD InStock
470349-618 470349-670
Class II Laser Pointers
Laser Pointers
Class 2 laser are safe for classroom use.
  • Maximum output of less than 1 mW
  • Push-button switch
These economical laser pointers are useful for many optical experiments. Their lower power output is ideal for middle school and high school classrooms. The lasers operate on a push-button switch, are housed in a rubberized aluminum pen-style case, and include a pocket clip.
The 6" long × 0.5" diameter Class 2 Red laser pointer has a maximum output of 1 mW and a wavelength of 650 nm (±10 nm). Range of beam is 75 meters. Operates on two 'AAA' batteries (included).
The 6" long × 0.5" diameter Class 2 Green laser pointer has a maximum output of 1 mW and a wavelength of 532 nm (±10 nm). Range of beam is 125 meters. Operates on two 'AAA' batteries (included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty