Would you like to log in again?
Heat Transfer Kit
Supplier: United Scientific Supplies
UNHTRF01
470349-628EA 33 USD InStock
470349-628
Heat Transfer Kit
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Accurately plot the thermal energy transferred by an aluminum bar.
  • Investigate heat transfer by conduction
  • Easy-to-read digital thermometers
  • Durable foam calorimeter cups with lids
The Heat Transfer Kit allows students to investigate the transfer of heat by conduction between two heat reservoirs at different temperatures and to calculate the quantities of heat involved in the process. The kit includes two plastic foam calorimeters with lids, a U-shaped aluminum bar which acts as a 'heat bridge', two digital thermometers for tracking the temperature changes in the masses of water held inside the calorimeters, and an activity guide.
The calorimeters hold up to 200 ml of liquid and the digital thermometers measure temperatures between 50 and 150 °C with a precision of 0.1 °C. The aluminum bridge has a cross sectional area of 122 mm² and is about 200 mm long between the liquid reservoirs (the exact length can vary, depending on the volumes of water used).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty