Would you like to log in again?
Economy Calorimeter
Supplier: United Scientific Supplies
UNECNCLR
470349-676EA 32.29 USD InStock
470349-676
Economy Calorimeter
Calorimeters
Easily determine the caloric energy in food.
  • Determine the caloric count of snack food
  • Holds the food item safely
  • Includes activity guide
The economy food calorimeter allows students to investigate the amount of energy released when food is burned. This involves using the burning energy of food to heat a vessel of water. The increase in temperature of the water represents the energy given off by the burning food. During the investigation students will learn how much energy is required to burn off some food items, and also how to read the caloric specification on food wrappers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty