Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chain Clamp

Supplier: United Scientific Supplies
UNCCLMP01
470349-702EA 35.15 USD InStock
470349-702
Chain Clamp
Clamps General Purpose Clamps
Support larger labware.

  • Chain length is 21"
  • Supports objects from 2 to 6.5" in diameter
  • Support rod 8.5" long and 0.46" diameter

This apparatus is useful for holding objects that are too large for standard clamps, such as glass distillation columns and odd-shaped glassware. Chain and support band are made of strong chrome-plated brass, and the support rod is constructed of stainless steel. Chain links bend to conform to the shape of the object being held. The tension spring locks chain links in place to firmly hold objects. Chain is 21" long and can be adjusted to support an apparatus from 2 to 6.5" in diameter. Support rod is 0.46" in diameter with a length of 8.5".

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty