Would you like to log in again?
Double Adjustable Burette Clamp, Stainless Steel
Supplier: United Scientific Supplies
UNDBBR01
470349-706EA 15.4 USD InStock
470349-706
Double Adjustable Burette Clamp, Stainless Steel
Clamps General Purpose Clamps
Hold two burettes at adjustable angles.
  • Stainless steel
  • Jaws with PVC coating can be swiveled
  • Securely holds two burettes
This versatile clamp features two separate sets of adjustable jaws for securing two burettes or any type of cylindrical tubes simultaneously. The PVC-coated jaws can swivel 90 degrees, so they can be next to each other or separated. The jaws are rounded to provide a better grip on round tubes, and can be swiveled and locked at any angle. Plated brass nuts enable easy positioning of the clamps.
The all-stainless steel jaws open from 15 to 45 mm (0.5 to 1.75"). The die-cast aluminum boss head attaches to rods up to 13 mm (0.5") Ext.Ø.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty