Would you like to log in again?
Single Burette Clamp, Stainless Steel
Supplier: United Scientific Supplies
UNSBR01
470349-738EA 15.4 USD InStock
470349-738
Single Burette Clamp, Stainless Steel
Clamps General Purpose Clamps
Stainless steel clamp with curved PVC-coated jaws.
  • Rotating jaw with PVC coating
  • With aluminum boss head clamp
Excellent for holding small objects. The PVC-coated jaws are rounded to provide a better grip on round tubes, and can be rotated and locked at any angle. Includes a die-cast aluminum boss head for easy attachment to a support rod.
The all-stainless steel jaw opens from 15 to 45 mm (0.5 to 1.75"). The die-cast aluminum boss head attaches to rods up to 13 mm (0.5") Ext.Ø.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty