Would you like to log in again?
Clear Plastic Magnifier, 3×/6×
PMD002
470349-748EA 1.2 USD InStock
470349-748
Clear Plastic Magnifier, 3×/6×
Magnifiers
Easy to use magnifier for all ages.
  • Clear plastic magnifier with 3× and 6× magnification lenses
  • 110 mm (4.5") long; 3× lens is 50 mm in diameter and 6× lens is 20 mm in diameter
Clear plastic magnifier is 110 mm (4.50") in length and features dual lenses of 3X and 6X magnification. 3× lens is 50 mm in diameter and 6× lens is 20 mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty