Would you like to log in again?
UniStore™ Reagent Bottles, Narrow Mouth, HDPE, 90 ml
Supplier: United Scientific Supplies
UniStore™
BNM090 BNM090-PK/6
470349-792PK 10.3 USD InStock
470349-792 470339-492
UniStore™ Reagent Bottles, Narrow Mouth, HDPE, 90 ml
Bottles Plastic Bottles
A narrow mouth HDPE bottle supplied with a lined polypropylene lid.
  • Narrow mouth reagent bottles with 90 ml capacity
  • Made from HDPE
  • Bottles are translucent, pliable, and strong
UniStore™ narrow mouth bottle has a capacity of 90 ml and a lined polypropylene lid. Constructed of high density polyethylene, the bottle is translucent, pliable, and has excellent strength. Leakproof bottle designed to meet TSA 3-1-1 guidelines for domestic and international travel.
Ordering information: Lined polypropylene caps included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty