Would you like to log in again?
Digital Thermometer
Supplier: United Scientific Supplies
UNDGT001
470349-810EA 17.2 USD InStock
470349-810
Digital Thermometer
Thermometers Digital Thermometers
Accurately take the temperatures of different solutions and see an easy to read digital read out.
  • C and F display selectable
  • Power save mode
  • Max./min. functionality
This digital thermometer has a temperature range of −50 to 300 °C (−58 to 572 °F), with an accuracy of 10 ±1 degree. The max./min. key displays the maximum and minimum temperature. The power saving mode turns off after 10 minutes. Includes one 1.5V (LR1130) battery. Overall length is 6.1" and the probe is 4" long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty