Would you like to log in again?
Multiparameter PH/EC/DO Turbidity Meter
Supplier: Hanna
HI9829-01041 HI9829-01041
470349-824EA 3738.39 USD InStock
470349-824 CA470349-824
Multiparameter PH/EC/DO Turbidity Meter
Electrochemical Meters pH and Multi-Parameter Meters
This multiparameter meter is a waterproof portable logging multiparameter meter. The microprocessor based multi-sensor probe allows for the measurement of key parameters including pH, ORP, conductivity, dissolved oxygen, turbidity, and temperature. The complete system is simple to setup and easy to use.
  • Color coded, field replaceable sensors
  • Great for environmental, field, and laboratory testing
  • Digital probe that is less than 2" in diameter
  • 3 color-coded sensor inputs
  • IP 68 rating for continuous immersion in water
  • Multistrand - multiconductor cable rated for 150 lbs. (intermittent use)
  • Weighted shield protects all sensors
  • Quick calibration - The quick calibration feature allows for a single point calibration for pH, conductivity, and dissolved oxygen all in few simple steps
Waterproof Protection - The water quality meter is enclosed in an IP 67 rated waterproof casing and can withstand immersion in water at a depth of 1 m for up to 30 minutes. The probe features an IP68 rating for continuous immersion in water Backlit Dot Matrix LCD Display - The meter features a backlit graphic LCD with on-screen help and the capability to display up to twelve parameters simultaneously. The graphic display allows for the use of virtual keys to provide for an intuitive user interface.
Intuitive Keypad - The fitted rubber keypad has dedicated keys for power, backlight, up/down arrows, help and alphanumeric characters. The meter also features two virtual soft keys that navigate the user through the configuration of each parameter, meter setup and logging of data. The interface is intuitive for any user’s level of experience. Dedicated Help Key - Contextual help is always available through a dedicated ‘HELP’ key. Clear tutorial messages and directions are available on-screen to guide users quickly and easily through setup and calibration. The help information displayed is relative to the setting/option being viewed.
Auto-sensor Recognition - The probe and meter automatically recognize the sensors that are connected. Any ports not used on the probe will not have the parameter displayed or be configurable
Automatic temperature compensation - Integrated temperature sensor allows for automatic temperature compensation of pH, conductivity, and dissolved oxygen measurements. Quick Calibration - Quick Calibration provides a speedy, single point calibration for pH, conductivity, and dissolved oxygen. Standard calibration options are available including pH up to three points, Conductivity at one point and dissolved oxygen up to two points.
Data Logging - This meter allows users to store up to 44,000 continuous or log-on-demand samples with logging intervals from one second to three hours. Rugged thermoformed carrying case - The HI9829 meter, probe, and all accessories are supplied in a rugged carrying case designed to provide years of use. The inside compartment of the carrying case is thermoformed to securely hold and protect all the components.
Ordering information: Replacement probes and sensors sold seperately. Calibration solutions sold seperately. pH storage and cleaning solutions sold seperately. Contact your sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty