Would you like to log in again?
Waterproof Pocket pH Tester
Supplier: Hanna
HI98107 HI98107
470349-826EA 58.89 USD InStock
CA470349-826 470349-826
Waterproof Pocket pH Tester
Electrochemical Meters pH and Multi-Parameter Meters
This pH tester is an easy-to-use pH water tester that makes monitoring water quality simple. This accurate pH tester features 0.1 resolution with automatic two-point calibration and temperature compensation in a single, portable, pocket device.
  • Compact design fits right in your pocket so you can take it anywhere
  • Cloth junction providing a larger surface area than a standard ceramic junction
  • Great for drinking water, aquariums, pools, and ponds
Features a large multi-level LCD which displays both pH and temperature readings simultaneously. The pH readings are displayed with a 0.1 pH resolution and with an accuracy of ±0.1 pH. The pH range of the HI98107 is from 0.0 to 14.0 pH. An exposed temperature sensor allows for rapid automatic temperature compensated pH measurements. Temperature can be set to display in ℃ or ℉. The LCD screen has stability and calibration tag indicators. The battery percent level is displayed at start up alerting the user to the remaining battery power that is available.
The pH electrode uses a unique reference cell design that utilizes a cloth junction that provides a larger surface area than a standard ceramic junction.
Calibration is performed by pressing the dedicated calibration button. Once in calibration mode the pH electrode can be calibrated to 1 or 2 points using the pre-programmed standard buffers (pH 4.01, 7.01, 10.01). When the standard value is recognized and stability is reached, the meter automatically accepts and stores the calibration value.
Setup options include automatic shut off timing and temperature unit displayed. Automatic shut-off options are to turn the water pH meter off after 8 minutes, 60 minutes, or disabled. To enter the setup mode the user presses a button located in the battery compartment.
Ordering information: If pH probe dies, will have to repurchase product. pH calibration solutions, storage solutions, and cleaning solutions sold seperately. Contact your sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty