Would you like to log in again?
HALO2 Wireless General-Use pH Tester
Supplier: Hanna
HI9810412 HI9810412
470349-828EA 161.51 USD InStock
CA470349-828 470349-828
HALO2 Wireless General-Use pH Tester
Electrochemical Meters pH and Multi-Parameter Meters
The HALO2 Wireless General Use pH Tester has a refillable glass body and spherical glass sensing bulb made for users that prefer a low-maintenance laboratory-grade pH electrode.
  • Open-source Bluetooth allows easy integration into current data logging systems
  • IP65 water resistant protection
  • Specialized electrode for small volumes
Cleaning and disinfection solution – Your HALO2 pH tester comes with cleaning solution that has been engineered to keep your electrode free from grease and debris. This concentrated combination of detergents cleans your meter quickly and effectively so that your meter stays in optimum condition.
Easy calibration – HALO2 can be placed right into our calibration buffer sachets for easy calibration. Using a Hanna calibration sachet ensures your buffer is always fresh.
Storage solution – Proper storage of your electrode will ensure reliable measurements and a long electrode life. Our storage solution is specially formulated to keep the pH glass hydrated while minimizing organic growth.
Ordering information: If pH probe dies, will have to repurchase product. pH calibration solutions, storage solutions, and cleaning solutions sold seperately. Contact your sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty