Would you like to log in again?
pH Electrode, PEI Body Field Application
Supplier: Hanna
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HI12300 HI12300
470349-830EA 147.03 USD InStock
CA470349-830 470349-830
pH Electrode, PEI Body Field Application
Electrodes pH Electrodes
This pH electrode is a plastic body, double junction, gel filled pH electrode with a built-in temperature sensor for temperature compensated measurements in a single probe design.
  • Spherical glass tip
  • Double junction reference
  • 3.5 mm connector
This probe features a 3.5 mm connector designed exclusively to be used with Hanna Instruments edge® Tablet Meters. It also features a single ceramic junction, and the spherical pH sensing portion is made with general purpose glass. The electrode has a rugged PEI (polyetherimide) body that has the chemical resistance of a glass body in an unbreakable plastic making it ideal for field and industrial applications.
PEI electrode body - The PEI (polyetherimide) electrode body is suitable for a wide range of applications and excels in field measurements due to its durability. The shielding around the spherical glass tip minimizes breakage due to accidental bumping or dropping of the electrode. The PEI plastic is a high-quality plastic that is chemically resistant to many harsh chemicals.
Ordering information: pH calibration and storage solutions sold seperately. Contact your sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty