Would you like to log in again?
High Range PH/EC/TDS Tester
Supplier: Hanna
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HI98130 HI98130
470349-836EA 199.99 USD InStock
470349-836 CA470349-836
High Range PH/EC/TDS Tester
Electrochemical Meters pH and Multi-Parameter Meters
Accurate. Waterproof. No more switching between meters for your routine measurements with the HI98130 pH and conductivity tester.
  • Compact and waterproof design fits right in your pocket so you can take it anywhere
  • Extractable cloth junction extends the life of your pH and conductivity tester
  • Great for hydroponics, aquariums, pools, and ponds
  • 3 sensors in a single meter
  • pH electrode with extractable junction
  • Amperometry EC/TDS sensor
  • Exposed temperature probe for faster EC/TDS readings
This multiparameter tester is a highly accurate, waterproof combo pH EC tester that can also measure EC/TDS in the high range (0.00 to 20.00 mS/cm EC/ 0.00 to 10.00 ppt TDS) and temperature. This floating, waterproof combination tester has an easy to read, LCD and an automatic shut-off. In addition, pH and EC/TDS readings are automatically temperature compensated (ATC) to prevent temperature-related variations in your measurements. For EC/TDS readings, the EC/TDS conversion factor (CONV) is selectable by the user, as well as the temperature compensation coefficient β (BETA).
Ordering information: Replaceable pH electrode is purchased seperately. pH calibration solutions, storage solutions, and cleaning solutions need to be purchased seperately. Contact your sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty