Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
TI-34 Multiview Calculator
Supplier: D&H
34MV/BK-A TI34MULTIVIEW TI34MULTVIEWTK
470348-870EA 279.89 USD InStock
470348-870 CA470157-974 470349-966
TI-34 Multiview Calculator
Calculators Pocket Calculators
Same functionality as the TI-34II Explorer plus display and feature enhancements.
  • Designed with educators and ideal for middle school math
  • MultiView display shows fractions as they are written on paper
  • 4-line display enables scrolling and editing of inputs
  • Simplify and convert fractions to decimals and back again
  • Integer division key expresses results as quotient and remainders
  • 6⁵/₈L×3³/₈W×⁷/₈"H Size
Features MultiViewTM Display, Up to 4 lines of Display, Fraction Simplification, Integer Division, Standard Math Notation, Toggle Key, Scrolling and Editing, User-Friendly Menus, Data/List Editor, Two Constant Operators, and Dual Power.
Ideal for algebra and trigonometry, this scientific calculator converts fractions to decimals and vice versa. The integer division key also expresses results as quotient and remainders. Additional features include an LCD screen that reads in four lines and 16 characters, seven memory variables, scientific notation, and both solar and battery power.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty