Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
TI-83 Plus Graphing Calculator
Supplier: D&H
TI83PLUS TI-83PLUS
470349-968EA 169.99 USD InStock
470349-968 CA470151-724
TI-83 Plus Graphing Calculator
Calculators Pocket Calculators
This calculator has enhanced graphing capabilities.
  • Graphing Capabilities
  • TI-83 Plus Compatible with the LabPro, CBL, and CBR 2
  • Eight-Line Display
  • Split Screen for Simultaneous Graph and Tabular Display
  • Functions for Finance, Algebra, Calculus, and More
The TI-83 features advanced functions for finance, algebra, calculus, statistics, and science. The display allows 16 characters per line with a maximum of eight lines and has a split screen for simultaneous graph and tabular display. It also contains Flash technology, so users can upgrade to future software sets or quickly store and recall large programs.
This calculator is compatible with the LabPro, CBL 2, and CBR; and the included unit-to-unit cable allows data sharing with another TI-82, 83, or 83 Plus calculator. It has 24K RAM of memory and 160K Flash ROM.
Ordering information: This calculator includes a user’s manual, four AAA batteries, and a backup CR1616 lithium battery. The teacher kit includes ten calculators, ten instruction booklets, a poster for quick reference, and calculator-to-calculator link cables.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty