Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
TI-30 Xa Scientific Calculator
Supplier: D&H
TI30XA
470349-970EA 17.59 USD InStock
470349-970
TI-30 Xa Scientific Calculator
Calculators Pocket Calculators
This fundamental calculator combines statistics and advanced scientific functions for high school math and science.
  • Adds, subtracts, multiplies, and divides fractions entered in traditional numerator/denominator format
  • Performs trigonometric functions, logarithms, roots, powers, reciprocals, and factorials
  • Polar/rectangular conversions
  • One-variable statistics include results for mean and standard deviation
  • Battery powered
This scientific calculator is an industry standard, and a workhorse in classrooms around the world. A solid solution for general math, algebra, trigonometry, and statistics.
Ordering information: This calculator includes a slide case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty