Would you like to log in again?
VOC/Formaldehyde Meter
Supplier: NEW GENIE GROUP INC SE
VFM200
470350-972EA 447.6 USD InStock
470350-972
VOC/Formaldehyde Meter
Electrochemical Meters Ion Meters
The VFM200 measures VOC (Volatile Organic Compound) and formaldehyde concentrations in real-time.
  • LCD display
  • Rechargeable battery
  • 0.01 ppm resolution
  • 5% accuracy
Fast response time, compact, easy-to-carry, and excellent for checking the indoor air quality.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty