Would you like to log in again?
nUVaClean™ UV Pipette Sterilizer
Supplier: MTC Bio
P5590 P5590-E
470350-978CS 198.9 USD InStock
470350-978 470350-980
nUVaClean™ UV Pipette Sterilizer
Pipette Stands
MTC’s nUVaClean™ is the first pipette rack that not only organizes and protects six pipettes, but also destroys greater than 99% of unwanted contamination.
  • UV-C lamp and reflector system destroys 99% of DNA
  • Eliminates cross-contamination
  • 360° UV exposure
  • Fits all major brands
  • Compact 6.5'' (17 cm) footprint
The new nUVaClean™ uses a germicidal UV-C lamp. A high-efficiency reflector/concentrator completely bathes pipette shafts in 254 nm UV light. This exposure destroys unwanted microbiological contaminants and prevents cross-contamination in PCR and other sensitive procedures. Just place single-channel pipettes of any range or brand in the nUVaClean™ and press the 'start' button to begin the auto-decon cycle. After 28 minutes the nUVaClean will power down automatically, delivering up to six pipettes that are safe, clean and ready for use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty