Would you like to log in again?
Celestron SkySync GPS Accessory
Supplier: CELESTRON INTERNATIONAL INC SE
Celestron
93969
470351-060EA 203.99 USD InStock
470351-060
Celestron SkySync GPS Accessory
Telescopes
Add GPS to your Celestron computerized telescope and get time, date, and location downloads from orbiting Global Positioning Satellites.
  • Upgrade your Celestron computerized telescope to GPS with SkySync
  • Improve the speed and accuracy of alignments with 16-channel GPS
  • Automatically updates the time and date
  • Plugs into the AUX port on the base of the telescope
  • Compatible for all non-GPS Celestron computerized telescopes
This 16-channel GPS module plugs into your telescope’s hand control or the auxiliary (AUX) port on your drive base and then mounts to a tripod leg to stay safely out of the way with included straps.
SkySync makes it easier to align your telescope while keeping your date and time updated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty