Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
ACCU-SCOPE® EXC-350 Upright Microscopes
Supplier: ACCU-SCOPE INC SE
ACCU-SCOPE®
EXC-350-SPH EXC-350-560 EXC-350-PHDF EXC-350-PH EXC-350 EXC-350-HDS EXC-350-470LP EXC-350-470 EXC-350-HDSLT EXC-350-SAPO
470351-118EA 1952 USD InStock
470351-118 470351-114 470351-224 470351-212 470351-176 470351-230 470351-112 470351-078 470351-260 470351-150
ACCU-SCOPE® EXC-350 Upright Microscopes
Microscopes
Delivers a high level of optical performance, versatility and durability for clinical, veterinary and education laboratory environments.
  • Trinocular viewing head
  • Digital microscope options
  • Infinity corrected
  • Inclined 30°
  • Optional fluorescence observation
  • 3 W LED with Variable Intensity Control
  • PL10×/20 mm focusable eyepiece with diopter for ease of use for the visually impaired
  • Reversed quintuple nosepiece with space to add objectives
  • Infinity corrected PLAN 4×, 10×, 40×R and 100×R oil objectives
  • Mechanical stage with low position coaxial controls, right hand control
With our infinity plan objectives and brilliant LED illumination, the EXC-350 produces bright, crisp, and sharp images. The trinocular port is ready to accept a microscopy camera (requires c-mount camera adapter) for documentation, analysis, and instructional purposes. The EXC-350 is available with an optional 1, 2, or 3-channel fluorescence accessory with integrated LED illumination. Applications for fluorescence observation include immunostaining, fungus and bacteria identification, nucleic acids and genetics, and protein localization.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty