Would you like to log in again?
ACCU-SCOPE® EXC-120 Upright Microscopes
Supplier: ACCU-SCOPE INC SE
ACCU-SCOPE®
EXC-121-V3 EXC-123-WIFI EXC-120-PL-RC EXC-121-3 EXC-121-VD3 EXC-123-HDS EXC-123-PL-RC EXC-120-3 EXC-123 EXC-120-RC EXC-123-PH EXC-123-PL EXC-121-VD EXC-120 EXC-121-V EXC-123-EC50 EXC-123-HD
470351-228EA 1343 USD InStock
470351-228 470351-218 470351-258 470351-154 470351-264 470351-164 470351-076 470351-142 470351-156 470351-194 470351-282 470351-086 470351-174 470351-162 470351-250 470351-272 470351-090
ACCU-SCOPE® EXC-120 Upright Microscopes
Microscopes
The EXC-120 Series is engineered for classroom, medical and veterinary applications.
  • Binocular, Monocular or Trinocular viewing head
  • Digital microscope options
  • Head is rotatable 360°
  • WF10×/18 mm eyepiece (paired - one with pointer)
  • Diopter adjustment for ease of use with the visually impaired
  • Reversed quadruple nosepiece
  • DIN 4×, 10×, SP40×R and 100×R oil objectives
  • Rackless mechanical stage
Delivers sharp, high contrast objectives, ergonomic operation, and a rugged, die-cast aluminum alloy frame, the EXC-120 series will deliver excellent performance through years of heavy use. ACCU-SCOPE® offers the EXC-120 series with the options of brightfield observation, darkfield observation, phase contrast, simple polarization, in a trinocular, binocular, monocular or dual view observation viewing head, and the option of adding a camera for your digital microscope options.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty