Would you like to log in again?
ACCU-SCOPE® EXC-100 Student Upright Microscopes
Supplier: ACCU-SCOPE INC SE
ACCU-SCOPE®
EXC-103 EXC-103-DF EXC-103-EC50 EXC-103-WIFI EXC-100 EXC-100-DF EXC-103-EP EXC-103-HDS EXC-103-HD
470351-170EA 625 USD InStock
470351-170 470351-192 470351-248 470351-116 470351-126 470351-186 470351-252 470351-084 470351-196
ACCU-SCOPE® EXC-100 Student Upright Microscopes
Microscopes
Engineered for classroom applications, the EXC-100 Series is built on a rugged, die-cast aluminum frame designed for years of use.
  • Binocular or trinocular microscope
  • Brightfield or Darkfield contrast method
  • WF10×/18 mm eyepieces (one with pointer)
  • 4×, 10×, 40×R and 100×R oil objectives
  • Optional camera and/or camera with monitor
  • Mechanical stage
  • N.A. 1.25 Abbe condenser on spiral mount with iris
  • Variable rechargeable LED illumination, with right angle power cord
  • Cord wrap
Featuring ergonomic focusing controls, all metal focusing gears and a wide field of view, the EXC-100 Series delivers an exceptional balance of value and performance in an educational microscope. Choose between binocular (EXC-100) and trinocular (EXC-103) viewing models.
Ideally suited for the classroom environment, the EXC-100 Series provides clear images during observation. with front lens surfaces that are flush and easy to clean. EXC-100 microscopes come standard with Achromat 4×, 10×, 40× and 100x oil immersion DIN objectives.
The economical LED illuminator in the EXC-100 Series uses much less energy than traditional microscopes, plus it is rechargeable, so the microscope can be operated with or without a cord. The low positioned coaxial focusing mechanism provides complete ergonomic comfort to users during extended viewing periods. The coaxial focusing controls allow the user to rest their hand on the counter while focusing the microscope, allowing for maximum comfort.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty