Would you like to log in again?
ACCU-SCOPE® EXI-310 Trinocular Inverted Microscopes
Supplier: ACCU-SCOPE INC SE
ACCU-SCOPE®
EXI-310-PL EXI-310-PH-HDS EXI-310-PH
470351-280EA 3032 USD InStock
470351-280 470351-200 470351-134
ACCU-SCOPE® EXI-310 Trinocular Inverted Microscopes
Microscopes
A trusted and dependable inverted microscope.
  • 5 W transmitted LED
  • Phase contrast and interference filter sliders
  • Extra-long working distance (ELWD) condenser
  • Clear glass stage plate
  • Optional attachable mechanical stage
  • Binocular tubes w/WF10×/22 mm eyepieces
  • Trinocular camera port
  • Fixed plain stage or optional attachable mechanical stage
  • Coaxial coarse and fine focus
  • Optional epi-fluorescence illuminator: GFP/FITC, Cherry Red, Texas Red, TRITC-Cy3,FITC/EGFP Longpass, YFP, DAPI, tDtomato Alexa 568
  • Optional 2 or 3 position fluorescence filter cube selector (required for fluorescence)
  • Shield for epi-fluorescence illuminator (standard with optional epi-fluorescence illuminator)
  • Quintuple nosepiece with long working distance objectives (selection of plan, plan phase, plan fluor, plan fluor phase)
  • Variable transmitted light intensity
Ideal for routine tissue culture, clinical laboratory observations and teaching applications, the EXI-310 is available in three standard configurations (phase contrast, 2-channel fluorescence, 3-channel fluorescence) or as a fully modular microscope to customize to meet your individual needs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty