Would you like to log in again?
ACCU-SCOPE® EXM-50 Student Monocular Microscope
Supplier: ACCU-SCOPE INC SE
ACCU-SCOPE®
EXM-50
470351-240EA 159 USD InStock
470351-240
ACCU-SCOPE® EXM-50 Student Monocular Microscope
Microscopes
Designed to provide educators with a student-ready instrument at an affordable price.
  • DIN 4×, 10×, 40×R objectives
  • Flat top stage with stage clips and disc diaphragm
  • Coarse and fine focusing control
  • Variable LED illuminator
  • 3500 mah NI rechargeable battery
A robust microscope that is ideal for the educational setting: compact footprint, built-in LED illumination, built-in carry handle, simple set-up and storage. Designed for elementary and middle school application, the EXM-50 offers DIN Achromat 4×, 10×, 40×R* objectives (* spring loaded).
The EXM-50 features an LED illumination system allowing corded and cordless operation. For cordless operation, simply insert AA alkaline batteries (not included) into the battery compartment on the bottom of the microscope. The EXM-50 is super portable thanks to its integrated carrying handle making it easier than ever for students and teachers to move the microscope around the classroom. The viewing head is rotatable 360° making the EXM-50 easy to store, too.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty