Would you like to log in again?
Estes Preassembled Rocketry Parachutes
Supplier: ESTES INDUSTRIES INC SE
Estes
2265 2268 2267 2264 2262 2271
470351-950EA 7.75 USD InStock
470351-950 470351-952 470351-928 470351-936 470351-958 470351-926
Estes Preassembled Rocketry Parachutes
Educational Materials Physics Educational Materials Mechanics Learning Activities
Lightweight, tear-resistant parachutes in a variety of sizes.
  • Blown film plastic is lightweight yet durable
  • Hexagonal shape, with shroud lines pre-attached in three loops
  • Shroud lines will not break in flight, nor fray over time
  • Multiple sizes for any sized rocket
These parachutes come preassembled and ready to use on your choice of model rockets. The shroud lines come attached, and it simply attaches to the nose cone by sliding the shroud lines through the anchor then looping the parachute through them. This lightweight but strong parachute is designed to fit into small airframes and bring the rocket down to the ground with a light touch. Perfect for hobbyists, or use them in the science classroom to study the effects of drag as a function of diameter!
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty