Would you like to log in again?
miniPCR® Starter Classroom Pack
Supplier: AMPLYUS LLC
miniPCR®
QP-2500-16
470352-068EA 4505.29 USD InStock
470352-068
miniPCR® Starter Classroom Pack
Educational Classroom Kits and Activities
Bring cutting-edge biotechnology into your classroom with the miniPCR® Starter classroom pack.
Ideal for classes between 16 and 32 students working in 4 to 8 lab groups.
Certifications: The miniPCR® thermal cyclers and blueGel electrophoresis systems are CE marked, made in the USA, and backed by the miniPCR 2-year Warranty and 30-day satisfaction guarantee!
Delivery information: This Starter Classroom Pack includes: 2 miniPCR® mini16X thermal cyclers: Featuring 16-well capacity, USB and Bluetooth connectivity, mini16 is broadly compatible with computers and smartphones including iPads and iPhones. 4 blueGel™ electrophoresis systems: Fast, durable, safe, and easy to use, blueGel™ allows you to visualize results instantly in a convenient, all-in-one device compatible with safe DNA dyes such as GelGreen®. 1 Gyro™ Microcentrifuge: This robust, single speed microcentrifuge is user-friendly and includes rotors for both 1.5 ml tubes and 0.2 ml tubes. Micropipetting supplies: Our H-style micropipettes are durable, accurate, and fully adjustable. Pack includes 4 student micropipettes (2 to 20 μl), 1 teacher set of 3 micropipettes (0.5 to 10 μl, 2 to 20 μl, 20 to 200 μl), and compatible tips (4 racks 200 μl tips, 2 racks 10 μl tips).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty