Would you like to log in again?
Unit™ Large Demonstration Set
Supplier: COCHRANES OF OXFORD LTD SE
Unit™
1962
470352-188EA 196.35 USD InStock
470352-188
Unit™ Large Demonstration Set
Educational Materials Chemistry Educational Materials Atomic Structure Learning Activities
Build models of organic and inorganic molecules that can be seen from the back of the class.
  • With large spheres 44 mm diameter and smaller spheres 25 mm for hydrogen
  • The versatile bonding peg system allows all the larger spheres to be joined in sp, sp2, sp3, dsp3 or d2sp3 configurations
  • Demonstrate single, double and triple bonds, isomers, ring structures, the chair-boat 'flip' of cyclohexane and VSEPR theory
This system offers great flexibility for teachers and lecturers. The models are easy to handle, build and take apart. The highly durable plastic parts create long-lasting models.
Packaging: Set Contents: 70 atom spheres, 90 bonds, bonding pegs and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty