Would you like to log in again?
Java Moss (Vesicularia dubyana)
Supplier: SEGREST FARMS
807-0020-9
470352-346EA 12.3 USD InStock
470352-346
Java Moss (Vesicularia dubyana)
Specimens Live Specimens Plants - Living Material
Give your aquarium an aged natural look.
  • Perfect for any size aquarium
  • Provide natural resting and hiding places for your smaller fish and invertebrates
  • Hardy and will survive in a wide range of aquarium parameters
Java moss is a popular aquascaping plants that grow on a variety of horizontal and vertical surfaces inside an aquarium. Can easily create a dense carpet on the bottom of your tank, giving it a colorful and natural appearance. For additional color in your habitat, and to help keep algae growth under control we recommend pairing with cherry shrimp.
Ordering information: Please specify a delivery date during checkout. Live moss will ship via overnight or second day delivery.
Packaging: Packed in a portion cup
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty