Would you like to log in again?
Innovating Science Unknown Compositions - Soda Ash
Innovating Science
IS2533
470352-534EA 4.15 USD InStock
470352-534
Innovating Science Unknown Compositions - Soda Ash
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Determine the purity of a soda ash sample via gravimetry and acid-base titration.
  • Contains enough materials needed for 25 groups
  • Teachers Manual and Student Study Guide copymasters are included
In this AP Chemistry experiment, students will compare two analytical methods: Gravimetry and Double Acid-Base Titration in this experiment by using these methods to determine the purity of sodium carbonate.
Ordering information: Includes: 10 Numbered Impure Soda Ash Samples, 25 g
Calcium Chloride, 75 g
2×500 ml Hydrochloric Acid Solution, 3 M
2×250 ml Sodium Hydroxide Standard Solution, 2 M
2×30 ml Bromothymol Blue Indicator
2×30 ml Methyl Orange Indicator
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty