Would you like to log in again?
Innovating Science Unknown Compositions - Ferrous Ammonium Sulfate
Supplier: ALDON CORP SE
Innovating Science
IS2532
470352-538EA 203 USD InStock
470352-538
Innovating Science Unknown Compositions - Ferrous Ammonium Sulfate
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Determine the purity of a ferrous ammonium sulfate sample via gravimetric and spectrophotometric analysis.
  • Contains enough materials needed for 25 groups
  • Teachers Manual and Student Study Guide copymasters are included
In this AP Chemistry experiment, students will perform Gravimetric Analysis and Spectrophotometric analysis to determine the purity of ferrous ammonium sulfate samples of unknown composition.
Ordering information: Inclues: 5 Numbered Impure Ferrous Ammonium Sulfate Samples, 25 g
5 Lettered Impure Ferrous Ammonium Sulfate Samples, 25 g
3×50 ml Ammonia Solution
Phenanthroline, 12.5 g
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty