Would you like to log in again?
biOrb Air 30
Supplier: GEOGLOBAL PARTNERS
85240 77669 72558 77668 71749 71748
470352-588EA 13.4 USD InStock
470352-588 470352-586 470352-580 470352-590 470352-584 470352-592
biOrb Air 30
Animal Supplies Animal Environmental Enrichment
The ultimate terrarium for plant enthusiasts.
  • Perfect lighting
  • Maintains humidity levels
  • Unique design and construction
  • Two year warranty
Showcase and grow healthy tropical, temperate, or semi-arid plants in this fully automated terrarium. The newest model, the AIR 30, features a compact size, making it ideal for desktop or tabletop display.
This terrarium senses when the humidity drops and gently mists and circulates air to stay within the selected relative humidity (RH) range. Use the included remote control to choose between three settings: low for semi-arid plants, moderate for temperate plants with no specific needs, and high for tropical plants. Natural daylight is not necessary as the lighting is arranged in an LED array at the top of the sphere. This emits the perfect light output that promotes natural growth and prevents plants from bending towards a single light source. Use the remote to control the brightness or pick an 8-hour, 10-hour, 12-hour, or 14-hour light cycle, with each cycle simulating sunrise, daylight, dusk, and nighttime.
Enjoy a 360 degree view of this mini paradise through its globe-like shape. Its acrylic material is 10 times stronger than glass, 50% lighter, and has a 23% transparency rating for a clear view. Plus, the low heat loss of acrylic makes it perfect for regulating the internal climate of the terrarium.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty