Would you like to log in again?
G.S.C. Projectile Launcher
Supplier: G.S.C. INTERNATIONAL, INC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PROLCH
470352-708EA 181.99 USD InStock
470352-708
G.S.C. Projectile Launcher
Educational Materials Physics Educational Materials Mechanics Learning Activities
Budget-friendly, yet reliable, this projectile launcher comes with plastic marbles for safety.
  • Includes two 16 mm plastic balls
  • Five different compression settings for the spring make the velocity highly adjustable
This Projectile Launcher can launch the plastic marbles that it comes with a maximum of 8 meters. The launch tube has five speed settings that can lock into place to assure consistent launches at the same velocities. There is a protractor printed on the base to measure the angle of launch. A photogate (not included) can be attached near the mouth of the launcher to accurately record initial velocity.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty