Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Digital Ammeter
Supplier: G.S.C. INTERNATIONAL, INC.
DAM-06
470352-712EA 43.79 USD InStock
470352-712
Digital Ammeter
Electrical Meters Ammeters
Perfect for a wide range of electrical projects and experiments.
  • Easy-to-read display
  • Powered by 9 V battery, included
Stop struggling with outdated analog meters or complex setups. This compact, battery-powered, digital device provides accurate readings from 0.00 to 19.99 A, making it perfect for a wide range of science, education, and DIY projects. With its easy-to-read display, you can quickly check the amps flowing through your circuits without any guesswork.
Simply connect it in series, and the digital ammeter will do the rest, powered by a simple 9 V battery. Whether you're a student, educator, or electronics enthusiast, this reliable ammeter is an essential tool to have on hand. Unlock the secrets of electricity, explore STEM concepts, or troubleshoot your latest creation - the digital ammeter puts the power of precise current measurement right at your fingertips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty