Would you like to log in again?
DC Motor with Alligator Leads
Supplier: GSC INTERNATIONAL SE
4-280LD
470352-720EA 11.69 USD InStock
470352-720
DC Motor with Alligator Leads
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The easiest small DC motor for your maker projects.
  • Insulated, color-coded leads with alligator clips
  • Simple to incorporate into any electronics project
This small, yet powerful motor is an ideal component for hundreds of electrical experiments and projects. The motor housing is 23.8 mm by 30.5 mm, with an overall length of 45 mm (including the 2 mm shaft). It operates on between 4.5 to 6 V. Each unit also incorporates two 30 mm long red and black connector cords with insulated alligator clips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty