Would you like to log in again?
Mechanical Wave Kit Parts and Accessories
Supplier: GSC INTERNATIONAL SE
WVGEN-STRING WVGEN-SPRING WVGEN-SAND
470352-702EA 10.29 USD InStock
470352-702 470352-722 470352-700
Mechanical Wave Kit Parts and Accessories
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Essential replacement parts for your mechanical wave investigations.
  • 40 grams of replacement blue sand for chladni plates
  • 0.5 mm thick×1 meter long string for demonstrating standing waves
  • Replacement spring has 0.75 mm thick wire, and measures 30 mm diameter×60 mm length (compressed)
Strings snap, springs deform, and sand gets swept into the garbage can. Keep your mechanical wave demonstrations ready to go with these high-quality replacement parts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty