Would you like to log in again?
Set of Three Laser Pointers
Supplier: GSC INTERNATIONAL SE
LSP-SET-3
470352-724EA 62.5 USD InStock
470352-724
Set of Three Laser Pointers
Laser Pointers
Perfect for exploring the science of wavelengths, refraction, and diffraction.
  • Three pen-style laser pointers: violet (approx. 445 nm), green (approx. 532 nm), and red (approx. 650 nm)
  • Class 3B lasers, 8.5 mW
  • Each laser is powered by 2 AAA batteries, included
This set is a handy tool for anyone interested in physics or optics. With three distinct wavelengths - violet, green, and red - you can explore the properties of different types of light. While laser pointers can be fun, it's crucial to use caution and avoid direct eye exposure.
The included batteries make this set ready to use right out of the box. Whether you're a student, teacher, or just curious about the science of light, this affordable laser pointer set is a great addition to your collection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty