Would you like to log in again?
10-Level Conductivity Indicators
Supplier: GSC INTERNATIONAL SE
CNDTEST-10 CNDTEST
470352-982EA 43.58 USD InStock
470352-982 470352-976
10-Level Conductivity Indicators
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
A very simple and visual demonstration of the level of conductivity of almost any solution.
  • Simple to use
  • Battery included and can be replaced as needed
  • LED Indicator light are a quick visual confirmation of the level of conductivity
  • Two pack sizes available
The 10-Level conductivity indicator can be used for quick comparisons of conductivity in solutions including, but not limited to, tap water, vinegar, baking soda solution, or varying concentrations of salt water. This device is very simple to use, just submerge both brass prongs in a solution hold the button on the side, and wait for the bar of LED's to display your results. Instruction are included.
Caution: Prop 65 Warning Label is listed on the package for lead and BPA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty