Would you like to log in again?
Butane Burner Hot Plate
Supplier: GSC INTERNATIONAL SE
BTN-PL-02
470352-978EA 83.92 USD InStock
470352-978
Butane Burner Hot Plate
Heaters Hot Plates
Unique, compact, and affordable staple for your lab!
  • The Butane Hotplate is has a low profile and wide base which provides a stable footing
  • The concentric rings support various sizes of flasks
  • The run time when full is approximately 25 minutes
The portable butane burner is a great option for any lab because the wide base and low profile provides the burner with a stable footing. The support rings attached at the top allow you to use the burner as a hotplate for laboratory glassware. The valve and ignition buttons are separate for added safety. The flame can reach a maximum temperature of approximately 1300 decrees Celsius. The burner can be refilled quickly and easily and holds approximately 30 grams of butane. The operating time is estimated to be 25 minutes. The product is shipped without fuel.
Caution: Prop 65 Warning Labels are applied for lead.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty