Would you like to log in again?
Human Fetal Development Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T251-15 T251
470353-126EA 12.35 USD InStock
470353-126 470353-092
Human Fetal Development Microslides
Slides Prepared Slides
Explore human development microscopically.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. Ova (left side 200×)/sperm (right 1800×) 2. Fourth day (left side - 200×)/sixth day - (right side 200×) 3. Fourth week (10×) 4. Thirty days (3×) 5. Seventh-eighth week (1/2×) 6. Fourth month (1.2×) 7. Five and one half months (1/5×) 8. Ninth month (1/4×).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty