Would you like to log in again?
The Cell Membrane Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T230-15 T230
470353-114BX 179.4 USD InStock
470353-114 470353-080
The Cell Membrane Microslides
Slides Prepared Slides
Explore the similarities and differences of various cell membranes.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects It on both sides
  • Set of 15 Includes detailed lesson pan on the topic under study
  • Durable, inexpensive teaching resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set Includes: 1. Epithelial cells (900×) 2. Cell membrane (3500×) 3. Paramecium-(dying) (200×) 4. Cilia -(Locomotion) (1500×) 5. Blood cells - (Structures) (26×) 6. Blood cells -(Functions) (600×) 7. White blood cell-(Recognition) (800×) 8. Onion skin cells (100×).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty