Would you like to log in again?
Taxonomy Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T217 T217-15
470353-134BX 179.4 USD InStock
470353-134 470353-122
Taxonomy Microslides
Slides Prepared Slides
Explore the classification systems of organisms.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. Bacteria in pairs - (1200×) 2. A spore producer - stained - (1000×) 3. Bacteria in chains - stained - (1000×) 4. Rod shaped bacteria - stained - (2000×) 5. Another spore producer - stained - (1000×) 6. Boils and abscess bacteria - stained - (1000×) 7. Syphilis bacterium - electron micrograph - (30000×) 8. Sauerkraut bacteria - stained - (2000×).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty